Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000904.hg.1 |
TC0X000066.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000904.hg.1 |
ENST00000380492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000904.hg.1 |
ENST00000482354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000904.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000904.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000497603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
ENST00000497603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145643.hg.1 |
TC0X00006671.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000380492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000460386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000461777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000482354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000495110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000497603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
ENST00000497603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_001177475.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006671.hg.1 |
NM_152581.3 |
Refseq |