Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038577.hg.1 |
TC02002452.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038577.hg.1 |
ENST00000264192.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038577.hg.1 |
ENST00000540637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038577.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200206839.hg.1 |
TC0200014693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000264192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000418920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000435117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000439355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000457793.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000462109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000483929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
ENST00000497432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014693.hg.1 |
NM_004288.4 |
Refseq |