Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036191.hg.1 |
TC03002044.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036191.hg.1 |
ENST00000273654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036191.hg.1 |
ENST00000442201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036191.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036192.hg.1 |
TC03002044.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036192.hg.1 |
ENST00000273654.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036192.hg.1 |
ENST00000442201.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036192.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209613.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209613.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209613.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209613.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209613.hg.1 |
TC0300014078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300209612.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209612.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209612.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209612.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209612.hg.1 |
TC0300014078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000442201.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000473854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
ENST00000489868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014078.hg.1 |
NM_181426.1 |
Refseq |