Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000328766.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000330154.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000356824.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000413506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000431597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000433318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000440675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000458024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000467416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000475854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000475854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000475854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000475854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000479333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000488982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000488982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000488982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000488982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000488982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000496581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000496581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000496581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
ENST00000496581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_001282231.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_014782.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010369.hg.1 |
NM_177949.3 |
Refseq |