Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080284.2 | NP_525023.2 | 35 - 36 | 4652 | 117 | |
XM_011524574.2 | XP_011522876.1 | 29 - 30 | 3980 | 117 | ||
XM_017024404.1 | XP_016879893.1 | 35 - 36 | 4964 | 117 | ||
XM_017024405.1 | XP_016879894.1 | 35 - 36 | 4964 | 117 | ||
XM_017024406.1 | XP_016879895.1 | 31 - 32 | 3932 | 117 | ||
XM_017024407.1 | XP_016879896.1 | 32 - 33 | 4036 | 117 | ||
XM_017024408.1 | XP_016879897.1 | 21 - 22 | 2525 | 117 | ||
GenBank mRNA | AF373290.1 | - | 34 - 35 | 4480 | 117 | |
AK097296.1 | - | 17 - 18 | 2079 | 117 | ||
AK125487.1 | - | 14 - 15 | 1744 | 117 | ||
AY028898.1 | - | 35 - 36 | 4652 | 117 | ||
U66680.1 | - | 6 - 7 | 669 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |