Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
ENST00000332811.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
ENST00000402174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
ENST00000462485.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
ENST00000544604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003476.hg.1 |
TC22000189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
TC2200009246.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168377.hg.1 |
NM_032173.3 |
Refseq |