Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261408.1 | NP_001248337.1 | 5 - 6 | 647 | 90 | |
NM_001261409.1 | NP_001248338.1 | 3 - 4 | 633 | 90 | ||
NM_003567.3 | NP_003558.1 | 3 - 4 | 610 | 90 | ||
XM_017002480.1 | XP_016857969.1 | 5 - 6 | 1023 | 90 | ||
GenBank mRNA | AF124250.1 | - | 3 - 4 | 602 | 90 | |
AK055442.1 | - | 3 - 4 | 633 | 90 | ||
BC028477.1 | - | 5 - 6 | 676 | 90 | ||
BC039895.1 | - | 3 - 4 | 503 | 6060541 | 90 | |
CN337133.1 | - | 3 - 4 | 612 | 90 | ||
U92715.1 | - | 3 - 4 | 418 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |