Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006708.hg.1 |
TC03000318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006708.hg.1 |
ENST00000266037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03006708.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153238.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153238.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153238.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153238.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153238.hg.1 |
TC0300007435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
ENST00000266037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007435.hg.1 |
NM_004947.4 |
Refseq |