Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022115.hg.1 |
TC01001413.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022115.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022115.hg.1 |
ENST00000484291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022115.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000490591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000490591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
ENST00000490591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176371.hg.1 |
TC0100010368.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176370.hg.1 |
TC0100010368.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000367943.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000463365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000464004.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000484291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000490591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000490591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
ENST00000490591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010368.hg.1 |
NM_007240.2 |
Refseq |