Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009371.hg.1 |
TC08000694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009371.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009371.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009371.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159349.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159349.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159349.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159349.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159349.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159349.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159349.hg.1 |
TC0800008658.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |