Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016948.hg.1 |
TC09001357.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016948.hg.1 |
ENST00000375326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016948.hg.1 |
ENST00000415431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016948.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016948.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173865.hg.1 |
TC0900010866.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000375326.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000414122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
ENST00000415431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_000507.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010866.hg.1 |
NM_001127628.1 |
Refseq |