Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028076.hg.1 |
ENST00000261653.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028076.hg.1 |
ENST00000392373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028076.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028076.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028076.hg.1 |
TC12002132.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
ENST00000540889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012451.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
ENST00000261653.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
ENST00000344271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
ENST00000392373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
NM_001980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
NM_194356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197394.hg.1 |
TC1200012451.hg.1 |
Clariom D TC |