Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000479789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000479789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000479789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000480580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000480580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
ENST00000480580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024930.hg.1 |
TC07001635.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024930.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024930.hg.1 |
ENST00000462558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024930.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000473569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000479789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000479789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000479789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000480580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000480580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
ENST00000480580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011905.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185185.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185185.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185185.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185185.hg.1 |
ENST00000005260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185185.hg.1 |
ENST00000462558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185185.hg.1 |
NM_018842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185185.hg.1 |
TC0700011905.hg.1 |
Clariom D TC |