Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009590.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009590.hg.1 |
ENST00000460338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009590.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009590.hg.1 |
TC13000855.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000486540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000486540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
ENST00000486540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164661.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164661.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164661.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164661.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164661.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164661.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164661.hg.1 |
TC1300009674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000376004.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000460338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000484943.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000486540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000486540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
ENST00000486540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009674.hg.1 |
NM_024089.2 |
Refseq |