Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008222.hg.1 |
TC02000526.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008222.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008222.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157691.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157691.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157691.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157691.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157691.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157691.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157691.hg.1 |
TC0200008300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |