Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001567.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001567.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001567.hg.1 |
TC21000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001566.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001566.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001566.hg.1 |
TC21000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
ENST00000538415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
ENST00000538415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007114.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148015.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148015.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148015.hg.1 |
TC2100007114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100148015.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148015.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148014.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148014.hg.1 |
NR_027695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148014.hg.1 |
TC2100007114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100148014.hg.1 |
ENST00000432141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148014.hg.1 |
NR_027695.1 |
Refseq |