Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001763.hg.1 |
TC6_apd_hap1000117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001763.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000390777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003595.hg.1 |
TC6_mcf_hap5000230.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003595.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003528.hg.1 |
TC6_ssto_hap7000219.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003528.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003956.hg.1 |
TC6_cox_hap2000232.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003956.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003770.hg.1 |
TC6_qbl_hap6000236.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003770.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003711.hg.1 |
TC6_dbb_hap3000209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003711.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003059.hg.1 |
TC6_mann_hap4000209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003059.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018814.hg.1 |
TC06004127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018814.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018814.hg.1 |
ENST00000446948.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018814.hg.1 |
NM_002118.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000390777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014276.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000414017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000418107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000438510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000477537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000487996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
ENST00000498020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201528.hg.1 |
TC0600014276.hg.1 |
Clariom D TC |