Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005514.hg.1 |
TC0X000410.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005514.hg.1 |
ENST00000276033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005514.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007655.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152586.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152586.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152586.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152586.hg.1 |
ENST00000587091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152586.hg.1 |
ENST00000590447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152586.hg.1 |
NM_006517.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152586.hg.1 |
TC0X00007655.hg.1 |
Clariom D TC |