Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020192.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020192.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020192.hg.1 |
TC10001576.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011574.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178833.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178833.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178833.hg.1 |
ENST00000260702.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178833.hg.1 |
NM_032211.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178833.hg.1 |
TC1000011574.hg.1 |
Clariom D TC |