Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014837.hg.1 |
ENST00000458411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010643.hg.1 |
ENST00000559380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013975.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013975.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013975.hg.1 |
TC22000826.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012631.hg.1 |
ENST00000514722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008199.hg.1 |
ENST00000605468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007390.hg.1 |
ENST00000463176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010069.hg.1 |
ENST00000591298.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163930.hg.1 |
TC2200008828.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200163930.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163930.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163930.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163930.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012631.hg.1 |
ENST00000514722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010643.hg.1 |
ENST00000559380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008199.hg.1 |
ENST00000605468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014837.hg.1 |
ENST00000458411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007390.hg.1 |
ENST00000463176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010069.hg.1 |
ENST00000591298.3 |
Ensembl |