Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301226.1 | NP_001288155.1 | 4 - 5 | 732 | 68 | |
NM_001301227.1 | NP_001288156.1 | 4 - 5 | 732 | 68 | ||
NM_003289.3 | NP_003280.2 | 4 - 5 | 732 | 68 | ||
NM_213674.1 | NP_998839.1 | 4 - 5 | 732 | 68 | ||
XM_017015087.1 | XP_016870576.1 | 4 - 5 | 604 | 68 | ||
XM_017015088.1 | XP_016870577.1 | 4 - 5 | 604 | 68 | ||
XM_017015089.1 | XP_016870578.1 | 4 - 5 | 604 | 68 | ||
XM_017015090.1 | XP_016870579.1 | 4 - 5 | 604 | 68 | ||
XM_017015091.1 | XP_016870580.1 | 4 - 5 | 604 | 68 | ||
XM_017015092.1 | XP_016870581.1 | 4 - 5 | 604 | 68 | ||
XM_017015093.1 | XP_016870582.1 | 4 - 5 | 604 | 68 | ||
GenBank mRNA | AK130045.1 | - | 4 - 5 | 320 | 68 | |
AK223258.1 | - | 4 - 5 | 601 | 68 | ||
AK294522.1 | - | 4 - 5 | 601 | 68 | ||
AK300249.1 | - | 3 - 4 | 696 | 68 | ||
AK304803.1 | - | 4 - 5 | 601 | 68 | ||
BC011776.2 | - | 4 - 5 | 552 | 3640927 | 68 | |
BE294047.1 | - | 3 - 4 | 293 | 68 | ||
EU106177.1 | - | 4 - 5 | 493 | 68 | ||
FJ224304.1 | - | 4 - 5 | 732 | 68 | ||
M12125.1 | - | 4 - 5 | 611 | 68 | ||
M74817.1 | - | 4 - 5 | 582 | 68 | ||
M75165.1 | - | 4 - 5 | 582 | 68 | ||
X06825.1 | - | 4 - 5 | 599 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |