Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000476343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000476343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000476343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000256496.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000419534.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000429403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000444332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000455168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000468010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000476343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000476343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000476343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
ENST00000611208.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006489.hg.1 |
NM_018184.2 |
Refseq |