Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080283.3 | NP_525022.2 | 27 - 28 | 3763 | 154 | |
XM_017024011.1 | XP_016879500.1 | 27 - 28 | 3728 | 154 | ||
XM_017024012.1 | XP_016879501.1 | 27 - 28 | 3832 | 154 | ||
XM_017024013.1 | XP_016879502.1 | 27 - 28 | 3652 | 154 | ||
XM_017024014.1 | XP_016879503.1 | 27 - 28 | 4252 | 154 | ||
XM_017024015.1 | XP_016879504.1 | 24 - 25 | 3275 | 154 | ||
GenBank mRNA | AF423307.1 | - | 26 - 27 | 3620 | 154 | |
AY028899.1 | - | 27 - 28 | 3763 | 154 | ||
CR627382.1 | - | 25 - 26 | 3456 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Homo sapiens Probe spans exons |