Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000338639.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000377493.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000460192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000465354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000469225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000493678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000497113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000497113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000497113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000497113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000497113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
ENST00000497113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_001123377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006729.hg.1 |
NM_007262.4 |
Refseq |