Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000508828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
ENST00000508828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013772.hg.1 |
TC03000758.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013772.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013772.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013772.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013766.hg.1 |
TC03000758.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013766.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013766.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013764.hg.1 |
TC03000758.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013764.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013764.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013765.hg.1 |
TC03000758.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013765.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013765.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164404.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164404.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164404.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164404.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164404.hg.1 |
TC0300008991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300164405.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164405.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164405.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164405.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164405.hg.1 |
TC0300008991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000507895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000508126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000508828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
ENST00000508828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008991.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164402.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164402.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164402.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164402.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164402.hg.1 |
TC0300008991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300164403.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164403.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164403.hg.1 |
ENST00000310546.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164403.hg.1 |
NM_080862.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164403.hg.1 |
TC0300008991.hg.1 |
Clariom D TC |