Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018375.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018375.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018375.hg.1 |
TC12001419.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000336399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000336399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000336399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000545609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000545609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018374.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018374.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018374.hg.1 |
TC12001419.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000336399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000336399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000336399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000545609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000545609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010497.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200179157.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179157.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179157.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200179157.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179157.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179157.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200179157.hg.1 |
TC1200010497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
ENST00000545609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
ENST00000256692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
ENST00000545609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
ENST00000550498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
NR_037144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200179160.hg.1 |
TC1200010497.hg.1 |
Clariom D TC |