Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023955.hg.1 |
ENST00000255082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023955.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023955.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023955.hg.1 |
TC11001994.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023954.hg.1 |
ENST00000255082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023954.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023954.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023954.hg.1 |
TC11001994.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011348.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187546.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187546.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187546.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187546.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187546.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187546.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187546.hg.1 |
TC1100011348.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100187547.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187547.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187547.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187547.hg.1 |
ENST00000255082.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187547.hg.1 |
ENST00000529256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187547.hg.1 |
NM_080658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187547.hg.1 |
TC1100011348.hg.1 |
Clariom D TC |