Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014786.3 | NP_055601.2 | 5 - 6 | 3687 | 58 | |
XM_017018623.1 | XP_016874112.1 | 5 - 6 | 2078 | 58 | ||
XM_017018624.1 | XP_016874113.1 | 4 - 5 | 1157 | 58 | ||
GenBank mRNA | AB002335.1 | - | 5 - 6 | 2461 | 58 | |
AF378754.1 | - | 5 - 6 | 3681 | 58 | ||
BC137228.1 | - | 5 - 6 | 3687 | 9021225 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |