Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003872.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003872.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003872.hg.1 |
TC16000261.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003870.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003870.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003870.hg.1 |
TC16000261.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003871.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003871.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003871.hg.1 |
TC16000261.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600152165.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152165.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152165.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152165.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152165.hg.1 |
TC1600007251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600152167.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152167.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152167.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152167.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152167.hg.1 |
TC1600007251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600152166.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152166.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152166.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152166.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152166.hg.1 |
TC1600007251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007251.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152168.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152168.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152168.hg.1 |
ENST00000005284.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152168.hg.1 |
NM_006539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152168.hg.1 |
TC1600007251.hg.1 |
Clariom D TC |