Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008917.hg.1 |
ENST00000302536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008917.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008917.hg.1 |
TC16000629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008916.hg.1 |
ENST00000302536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008916.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008916.hg.1 |
TC16000629.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600163621.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163621.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163621.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600163621.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163621.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163621.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600163621.hg.1 |
TC1600008498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000302536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
ENST00000563850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008498.hg.1 |
NM_020927.2 |
Refseq |