Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004468.hg.1 |
ENST00000257197.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004468.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004468.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004468.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004468.hg.1 |
TC18000437.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
TC1800008387.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157164.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257197.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
ENST00000257198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_004948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008387.hg.1 |
NM_024421.2 |
Refseq |