Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002336.2 | NP_002327.2 | 19 - 20 | 4222 | 63 | |
XM_006719078.3 | XP_006719141.1 | 19 - 20 | 4366 | 63 | ||
XM_011520671.2 | XP_011518973.1 | 18 - 19 | 3972 | 63 | ||
XM_017019304.1 | XP_016874793.1 | 17 - 18 | 4040 | 63 | ||
GenBank mRNA | AB209683.1 | - | 18 - 19 | 3674 | 63 | |
AF074264.1 | - | 19 - 20 | 4157 | 63 | ||
AK074543.1 | - | 4 - 5 | 590 | 63 | ||
AK074902.1 | - | 9 - 10 | 1759 | 63 | ||
BC117136.1 | - | 19 - 20 | 4114 | 40125687 | 63 | |
BC126405.1 | - | 19 - 20 | 4114 | 8992121 | 63 | |
BC143725.1 | - | 19 - 20 | 3979 | 9052239 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |