Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006901.3 | NP_008832.2 | 18 - 19 | 3053 | 110 | |
XM_006720539.2 | XP_006720602.1 | 18 - 19 | 3053 | 110 | ||
XM_011521613.2 | XP_011519915.1 | 18 - 19 | 3053 | 110 | ||
XM_011521614.2 | XP_011519916.1 | 19 - 20 | 2739 | 110 | ||
XM_011521615.2 | XP_011519917.1 | 18 - 19 | 2906 | 110 | ||
XM_011521616.2 | XP_011519918.1 | 18 - 19 | 2700 | 110 | ||
XM_011521617.2 | XP_011519919.1 | 18 - 19 | 3053 | 110 | ||
XM_011521618.2 | XP_011519920.1 | 18 - 19 | 3053 | 110 | ||
XM_011521619.2 | XP_011519921.1 | 17 - 18 | 2996 | 110 | ||
XM_011521620.2 | XP_011519922.1 | 17 - 18 | 2996 | 110 | ||
XM_011521621.2 | XP_011519923.1 | 17 - 18 | 2933 | 110 | ||
XM_011521622.2 | XP_011519924.1 | 18 - 19 | 3053 | 110 | ||
XM_017022228.1 | XP_016877717.1 | 18 - 19 | 2682 | 110 | ||
XM_017022229.1 | XP_016877718.1 | 19 - 20 | 2888 | 110 | ||
XM_017022230.1 | XP_016877719.1 | 17 - 18 | 2996 | 110 | ||
GenBank mRNA | AB209841.1 | - | 16 - 17 | 2431 | 110 | |
AB290183.1 | - | 17 - 18 | 2488 | 110 | ||
AF117888.1 | - | 18 - 19 | 2804 | 110 | ||
BC140869.1 | - | 18 - 19 | 2722 | 9021737 | 110 | |
BC150294.1 | - | 16 - 17 | 2061 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |