Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027100.hg.1 |
ENST00000265836.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027100.hg.1 |
ENST00000375682.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027100.hg.1 |
ENST00000429869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027100.hg.1 |
ENST00000525071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027100.hg.1 |
ENST00000525815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027100.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027100.hg.1 |
TC11002262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000524991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012219.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000375682.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000525071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000525815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000532513.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
ENST00000533664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
NM_017515.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193964.hg.1 |
TC1100012219.hg.1 |
Clariom D TC |