Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000586910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000586910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000586910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000253109.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000586910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000586910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000586910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000589181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
ENST00000592641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009607.hg.1 |
NM_031917.2 |
Refseq |