Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000264159.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000401392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000403017.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000412849.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000495945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000536680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000619650.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
NM_001286568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
NM_001286569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
NM_032143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000264159.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000401392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000403017.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000412849.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000495945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000536680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
ENST00000619650.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
NM_001286568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
NM_001286569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
NM_032143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204924.hg.1 |
TC0200014390.hg.1 |
Clariom D TC |