Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
TC02001415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
ENST00000347464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
ENST00000373525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
ENST00000392017.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
ENST00000392018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
ENST00000392020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
ENST00000498620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
NM_001190266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
NM_001190267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
NM_017974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
NM_030803.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02022525.hg.1 |
NM_198890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000347464.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000373525.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392017.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392020.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392021.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000417017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000419681.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000444735.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000474148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000479942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000498620.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_001190266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_001190267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_017974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_030803.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_198890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000347464.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000373525.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392017.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392020.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000392021.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000417017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000419681.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000444735.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000474148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000479942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
ENST00000498620.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_001190266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_001190267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_017974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_030803.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
NM_198890.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200180200.hg.1 |
TC0200011125.hg.1 |
Clariom D TC |