Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000349937.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000352065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000355717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000355995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000369395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000369397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000534894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000536810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000538897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000542695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000543371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
ENST00000545257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001146274.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001146283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001146284.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001146285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001146286.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001198525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001198526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001198527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001198528.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001198529.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_001198531.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
NM_030756.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009843.hg.1 |
TC10000820.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000346198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000349937.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000352065.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000355717.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000355995.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000369395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000369397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000536810.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000538897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000542695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000543371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000545257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000627217.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000629706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198528.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_030756.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000346198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000349937.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000352065.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000355717.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000355995.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000369395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000369397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000536810.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000538897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000542695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000543371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000545257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000627217.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
ENST00000629706.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146284.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001146286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198528.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_001198531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
NM_030756.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161138.hg.1 |
TC1000008942.hg.1 |
Clariom D TC |