Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001890.1 | NP_001881.1 | 12 - 13 | 384 | 103 | |
XM_006714089.2 | XP_006714152.1 | 12 - 13 | 400 | 103 | ||
XM_006714090.2 | XP_006714153.1 | 11 - 12 | 379 | 103 | ||
XM_006714091.2 | XP_006714154.1 | 11 - 12 | 376 | 103 | ||
GenBank mRNA | BC128227.1 | - | 12 - 13 | 373 | 40114618 | 103 |
BC128228.1 | - | 12 - 13 | 373 | 40114622 | 103 | |
U23157.1 | - | 12 - 13 | 363 | 103 | ||
X78416.1 | - | 12 - 13 | 384 | 103 | ||
X98084.1 | - | 11 - 12 | 312 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |