Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
TC04000374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
ENST00000246891.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
ENST00000354865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
ENST00000444405.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
ENST00000505782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
ENST00000507763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
ENST00000507772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005075.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000633258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000633258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
ENST00000633258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152917.hg.1 |
TC0400007769.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000246891.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000505782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000507772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000510936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000633258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000633258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
ENST00000633258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001025104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007769.hg.1 |
NM_001890.1 |
Refseq |