Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029346.hg.1 |
TC06002342.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029346.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029346.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
ENST00000630500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014031.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196698.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196698.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196698.hg.1 |
ENST00000366756.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196698.hg.1 |
NM_005618.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196698.hg.1 |
TC0600014031.hg.1 |
Clariom D TC |