Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012202.hg.1 |
ENST00000345988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012202.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012202.hg.1 |
TC16000838.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000588556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009252.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000345988.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000589090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000590782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
ENST00000592772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
NM_002705.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171101.hg.1 |
TC1600009252.hg.1 |
Clariom D TC |