Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000469584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011378.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000295899.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000464327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000469153.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000473141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000487570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
NM_025075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185327.hg.1 |
TC0300011378.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185331.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185331.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185331.hg.1 |
ENST00000498422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185331.hg.1 |
NM_001285404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185331.hg.1 |
TC0300011378.hg.1 |
Clariom D TC |