Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690580 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690608 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090880 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16427767 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690591 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690596 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690590 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090899 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16427760 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090886 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090906 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690579 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16427764 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690593 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090908 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090914 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090879 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16427769 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16427765 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16427771 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090917 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090881 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090911 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090915 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090877 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090907 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090916 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16427770 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690576 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090888 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P07690581 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090905 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090897 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090882 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090913 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28090883 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
33115 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AJ292259.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AJ292260.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AJ292261.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AJ292262.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AJ292263.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AJ292264.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AJ292265.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AK092392.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
AX747518.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
NM_025260.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
NM_138272.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
NM_138273.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
NM_138274.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
NM_138275.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
148728 |
NM_138277.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
99645 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
27113 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
42844 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
2083 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
880606 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P12896 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3386364 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P12896 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
25605 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P131840 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P122657 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P102029 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P131840 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P102029 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P122657 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
46147 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
53520 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
37743 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
28074 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P19178176 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P39294666 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P12896 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3386364 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3307073 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3386364 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P12896 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090904 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090911 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090899 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427760 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090888 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090918 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690591 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090917 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427764 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090905 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090915 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090913 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090906 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690590 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427771 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690603 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690593 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090897 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090914 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090886 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690596 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090907 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090897 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090886 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427764 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090917 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690596 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090907 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690590 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090906 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090899 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690591 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427771 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090911 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090918 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090904 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090905 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090888 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690603 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090913 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16427760 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07690593 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090915 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28090914 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P12896 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P122657 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P131840 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P102029 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690597 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690600 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690590 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690611 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690609 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690589 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690602 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690603 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690592 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690595 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690605 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690601 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690594 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690598 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690608 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P07690591 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16427767 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P07690595 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16427769 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16427768 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P07690597 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P07690592 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16427770 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P07690593 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P07690594 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P07690596 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P131840 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P102029 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P122657 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P122657 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P131840 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P102029 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P12896 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
80272 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
AB209653.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
AK092834.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
AK222857.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
AK309945.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
BC007339.2 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
CR602571.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
CR610479.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
CR611371.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
CR620554.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
CR621782.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
CR624957.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
NM_005794.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
106534 |
U31875.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
63453 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P102029 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P122657 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P131840 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
2738319 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
2813588 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
2738319 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
2813588 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690586 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690577 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16427759 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16427764 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16427767 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690609 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690600 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690605 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690582 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16427773 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16427768 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690611 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690601 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690584 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690591 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690593 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16427771 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690576 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690608 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690596 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690581 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690587 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690590 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16427761 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690594 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690579 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690589 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07690603 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P102029 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P122657 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P131840 |
- |
- |