Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010689.hg.1 |
TC04000775.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010689.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010693.hg.1 |
TC04000775.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010693.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010692.hg.1 |
TC04000775.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010692.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162106.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162106.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162106.hg.1 |
TC0400009048.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400162101.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162101.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162101.hg.1 |
TC0400009048.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400162107.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162107.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162107.hg.1 |
TC0400009048.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |