Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000566.hg.1 |
TC07000037.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000566.hg.1 |
ENST00000312371.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000566.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000566.hg.1 |
ENST00000489665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000566.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013329.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196203.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196203.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196203.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196203.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196203.hg.1 |
TC0700013329.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000312371.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000407112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000480560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
ENST00000485540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
NM_001284355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
NM_133463.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196208.hg.1 |
TC0700013329.hg.1 |
Clariom D TC |