Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000167.hg.1 |
TC09000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000167.hg.1 |
ENST00000382276.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000167.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000163.hg.1 |
TC09000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000163.hg.1 |
ENST00000382276.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000163.hg.1 |
ENST00000451501.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000163.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000162.hg.1 |
TC09000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000162.hg.1 |
ENST00000382276.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000162.hg.1 |
ENST00000451501.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000162.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144286.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144286.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144286.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144286.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144286.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144286.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144286.hg.1 |
TC0900006451.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900144287.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144287.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144287.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144287.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144287.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144287.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144287.hg.1 |
TC0900006451.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900144285.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144285.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144285.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144285.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144285.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144285.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144285.hg.1 |
TC0900006451.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144289.hg.1 |
TC0900006451.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000382276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000564322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
ENST00000569227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006451.hg.1 |
NM_021951.2 |
Refseq |