Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008213.1 | NP_001008214.1 | 3 - 4 | 428 | 99 | |
GenBank mRNA | BC032762.1 | - | 3 - 4 | 348 | 3457195 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |