Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000301336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000570858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000573398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000573398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000573398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000573398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000573398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
ENST00000574810.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009393.hg.1 |
NM_031430.2 |
Refseq |