Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001534.hg.1 |
TC09000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001534.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001534.hg.1 |
ENST00000379723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001534.hg.1 |
ENST00000379725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001534.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147256.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147256.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147256.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147256.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147256.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147256.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147256.hg.1 |
TC0900006968.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379721.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
ENST00000379725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006968.hg.1 |
NM_014471.2 |
Refseq |